Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK3 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4785JF525
Description
GRK3 Polyclonal antibody specifically detects GRK3 in Human samples. It is validated for Western BlotSpecifications
GRK3 | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 525 | |
adrenergic, beta, receptor kinase 2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
E. coli-derived recombinant human GRK3, Pro469-Leu688, Accession # P35626 | |
0.1 mL | |
Protein Kinase | |
157 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction