Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK3 Antibody (SR2232), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321869100UL
Description
GRK3 Monoclonal antibody specifically detects GRK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
GRK3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR2232 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
adrenergic, beta, receptor kinase 2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
A synthesized peptide derived from human GRK3 (Uniprot #: P35626) | |
100 μg | |
Protein Kinase | |
157 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction