Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK3 Rabbit anti-Human, Clone: 10O6S3, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31677220UL
This item is not returnable.
View return policy
Description
GRK3 Monoclonal antibody specifically detects GRK3 in Human samples. It is validated for Western BlotSpecifications
GRK3 | |
Monoclonal | |
Unconjugated | |
adrenergic, beta, receptor kinase 2, BARK2beta-adrenergic receptor kinase 2, beta-ARK-2, EC 2.7.11, EC 2.7.11.15, G-protein-coupled receptor kinase 3, GRK3Beta-ARK-2 | |
A synthetic peptide corresponding to a sequence within amino acids 589-688 of human GRK3 (P35626). | |
20 μg | |
Protein Kinase | |
157 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
10O6S3 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction