Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRK3 Recombinant Rabbit Monoclonal Antibody (10O6S3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542913
This item is not returnable.
View return policy
Description
Positive test controls include: Jurkat, SH-SY5Y. Immunogen sequence: EWRGEGESRQ NLLTMEQILS VEETQIKDKK CILFRIKGGK QFVLQCESDP EFVQWKKELN ETFKEAQRLL RRAPKFLNKP RSGTVELPKP SLCHRNSNGL.
ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase that is thought to act as a regulator of receptor function. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
Specifications
GRK3 | |
Recombinant Monoclonal | |
0.45 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35626 | |
GRK3 | |
A synthetic peptide corresponding to a sequence within amino acids 589-688 of human GRK3 (P35626). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
10O6S3 | |
Unconjugated | |
GRK3 | |
4833444A01Rik; ADRBK2; Adrbk-2; adrenergic receptor kinase, beta 2; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; AI851927; AW551196; BARK2; Bark-2; beta ARK2; beta-adrenergic receptor kinase 2; Beta-ARK-2; G protein-coupled receptor kinase 3; G-protein-coupled receptor kinase 3; Grk3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
157 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction