Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK4 Antibody (OTI4F5), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270845G
Description
GRK4 Monoclonal specifically detects GRK4 in Human samples. It is validated for Western Blot.Specifications
GRK4 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GRK4 | |
Recombinant protein fragment corresponding to amino acids 1-245 of human GRK4 (NP_892027) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI4F5 | |
Western Blot | |
EC 2.7.11, EC 2.7.11.16, G protein-coupled receptor kinase 2-like (Drosophila), G protein-coupled receptor kinase 4, G protein-coupled receptor kinase GRK4, GPRK2LIT11, GPRK4GRK4a, G-protein coupled receptor kinase 4, ITI1 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
2868.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction