Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK4 Antibody (OTI4F5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245914
Description
Ensure accurate, reproducible results in Western Blot
GRK4 Monoclonal specifically detects GRK4 in Human samples. It is validated for Western Blot.Specifications
GRK4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
EC 2.7.11, EC 2.7.11.16, G protein-coupled receptor kinase 2-like (Drosophila), G protein-coupled receptor kinase 4, G protein-coupled receptor kinase GRK4, GPRK2LIT11, GPRK4GRK4a, G-protein coupled receptor kinase 4, ITI1 | |
Mouse | |
66.4 kDa | |
0.1 mL | |
Protein Kinase | |
2868 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI4F5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
GRK4 | |
Human recombinant protein fragment corresponding to amino acids 1-245 of human GRK4(NP_892027) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction