Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK4 Monoclonal Antibody (OTI4F5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA805974

Description
This gene encodes a member of the guanine nucleotide-binding protein-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating its deactivation. This gene has been linked to both genetic and acquired hypertension.Specifications
GRK4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P32298 | |
GRK4 | |
Human recombit protein fragment corresponding to amino acids 1-245 of human GRK4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4F5 | |
Unconjugated | |
GRK4 | |
GPRK2L, GPRK4, GRK4a, IT11 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2868 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction