Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GRK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563388
Description
Immunogen sequence: ESGCFKDINK SESEEALPLD LDKNIHTPVS RPNRGFFYRL FRRGGCLTMV PSEKEVEPKQ Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 50%.
GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK4 phosphorylates the activated forms of G protein-coupled receptors. GRK4 has been linked to both genetic and acquired hypertension.
Specifications
GRK4 | |
Polyclonal | |
Unconjugated | |
GRK4 | |
G protein-coupled receptor kinase 4; G protein-coupled receptor kinase GRK4; GPRK2L; GPRK4; GRK4; GRK4a; IT11; ITI1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2868 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P32298 | |
GRK4 | |
Recombinant protein corresponding to Human GRK4. Recombinant protein control fragment (Product #RP-103285). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction