Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565030 Supplier Invitrogen™ Supplier No. PA565030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKQPIGRRLFR QFCDTKPTLK RHIEFLDAVA EYEVADDEDR SDCGLSILDR FFNDKLAAPL PEIPPDVVTE CRLGLKEENP SKKVFEECTR VAHNYLRGEP FEEYQESSYF SQFLQWKWLE R Highest antigen sequence identity to the following orthologs - mouse 66%, rat 61%.

GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK4 phosphorylates the activated forms of G protein-coupled receptors. GRK4 has been linked to both genetic and acquired hypertension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRK4
Gene Accession No. P32298
Gene Alias G protein-coupled receptor kinase 4; G protein-coupled receptor kinase GRK4; GPRK2L; GPRK4; GRK4; GRK4a; IT11; ITI1
Gene Symbols GRK4
Host Species Rabbit
Immunogen Recombinant Human GRK4. Recombinant protein control fragment (Product #RP-104593).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2868
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.