Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRK5 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299632APC
Description
GRK5 Polyclonal antibody specifically detects GRK5 in Human samples. It is validated for ELISASpecifications
GRK5 | |
Polyclonal | |
ELISA | |
EC 2.7.11, EC 2.7.11.16, FLJ39780, G protein-coupled receptor kinase 5, G protein-coupled receptor kinase GRK5, GPRK5FP2025 | |
Produced in rabbits immunized with purified, recombinant Human GRK5 (Accession#: NP_005299.1; Met 1-Ser 590) | |
0.1 mL | |
Protein Kinase | |
2869 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction