Learn More
Promega GRK5 Kinase Enzyme System, 10microg
Catalog No. PRV3981 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3981 10 μg
1 options
Catalog No. PRV3981 Supplier Promega Supplier No. V3981
Recombinant full-length human GRK5 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. GRK5 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family.
  • Easily Screen and Profile GRK5 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C