Learn More
Invitrogen™ GRK5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596262
Description
Immunogen sequence: LGCLIYEMIE GQSPFRGRKE KVKREEVDRR VLETEEVYSH KFSEEAKSIC KMLLTKDAKQ RLGCQEEGAA EVKRHPFFRN MNFKRLEAGM LDPPFVPDPR AVYCKDVLDI EQFSTVKGVN LDHTDDDFYS KFSTGSVSIP WQNEMIETEC FKELNVFGPN GTLPPDLNRN HPPEPPKKGL LQRLFKRQHQ NNSKSSPSSK TSFNHHINSN HVSSNSTGSS; Positive Samples: A-549, U-251MG, HepG2, 293T, HT-1080, Mouse liver, Mouse lung; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Peripheral membrane protein.
GRK5 is a serine/threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
Specifications
GRK5 | |
Polyclonal | |
Unconjugated | |
GRK5 | |
1200003L19Rik; Alpha-CP4; AlphaCP-4; FP2025; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; Gprk5; GRK; Grk5; GRK5 protein kinase; GRK-pan; Pan-GRK; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11476, 14773, 236300, 2869, 59075, 59092 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.96 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P34947, Q62833, Q8VEB1 | |
GRK5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 371-590 of human GRK5 (NP_0052991). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.