Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GRM2 (Human) Recombinant Protein
Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg
Description
Human GRM2 full-length ORF (NP_000830.2) recombinant protein without tag.
Specifications
Specifications
For Use With (Application) | Antibody Production |
Gene ID (Entrez) | 2912 |
Name | glutamate receptor, metabotropic 2 |
Quantity | 10 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | GLUR2, GPRC1B, MGLUR2, mGlu2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction