Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRM2 Mouse anti-Human, Clone:4A10B9, Abnova™

Catalog No. 89334021 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-021 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-021 Supplier Abnova Corporation Supplier No. MAB17597
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human GRM2.

L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen GRM2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A10B9
Conjugate Unconjugated
Description glutamate receptor, metabotropic 2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
Immunohistochemistry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene GRM2
Gene Alias GLUR2, GPRC1B, MGLUR2, mGlu2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 414-558 of human GRM2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2912
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.