Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GroEL Antibody (9A1/2), Novus Biologicals™
SDP

Catalog No. NB018822 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018822 0.025 mg
NBP19789505 0.05 mg
NBP19789502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018822 Supplier Novus Biologicals Supplier No. NBP1978950.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

GroEL Monoclonal specifically detects GroEL in E. coli samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen GroEL
Applications Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9A1/2
Conjugate Unconjugated
Dilution Western Blot 1:10000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Alias Cpn60 chaperonin GroEL, large subunit of GroESL, ECK4137, groEL, JW4103, mopA
Host Species Mouse
Immunogen E. coli GroEL.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948665
Test Specificity Recognizes Bacteria GroEL.
Target Species E. coli
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.