Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ groL Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144463
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144463 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144463 Supplier Invitrogen™ Supplier No. PA5144463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

groL Polyclonal Antibody for ELISA, Western Blot

Together with its co-chaperonin GroES, plays an essential role in assisting protein folding. The GroEL-GroES system forms a nano-cage that allows encapsulation of the non-native substrate proteins and provides a physical environment optimized to promote and accelerate protein folding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen groL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene groEL
Gene Accession No. P19421
Gene Alias groL; htpB; mopA
Gene Symbols groEL
Host Species Rabbit
Immunogen Recombinant Coxiella burnetii (strain RSA 493/Nine Mile phase I) CH60 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1209629
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.