Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone 2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299112MFV500
Description
Growth Hormone 2 Polyclonal antibody specifically detects Growth Hormone 2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Growth Hormone 2 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
GHL, GHVgrowth hormone variant, GH-VPlacenta-specific growth hormone, growth hormone 2hGH-V, placental-specific growth hormone | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Growth Hormone 2. | |
0.1 mL | |
Cytokine Research | |
2689 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction