Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone Antibody (5E9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Growth Hormone |
---|---|
Clone | 5E9 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Growth Hormone Monoclonal antibody specifically detects Growth Hormone in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Growth Hormone | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N | |
A synthesized peptide derived from Human Growth Hormone [UniProt P01241] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5E9 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2688 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title