Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone Monoclonal Antibody (KT19), Invitrogen™
Rat Monoclonal Antibody
Supplier: Thermo Scientific MA549587

Description
This product was used in both Indirect and Sandwich ELISA.
Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
Specifications
Growth Hormone | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P01241 | |
Gh1 | |
Recombinant Human GH1 | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
KT19 | |
Unconjugated | |
Gh1 | |
dog growth hormone, GH, Gh1, GHB1, GHB5, GHN, GH-N, Growth hormone, growth hormone 1, growth hormone 12, growth hormone B1, growth hormone B5, growth hormone precursor, HGH, hGH-N, IGHD1B, pituitary growth hormone, RNGHGP, Somatotropin | |
Rat | |
Protein A | |
RUO | |
2688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction