Learn More
Invitrogen™ Growth Hormone Polyclonal Antibody

Description
Purity is > 95% by SDS-PAGE.
Specifications
Specifications
| Antigen | Growth Hormone |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Gh1 |
| Gene Accession No. | P01241, P01244, P06880 |
| Gene Alias | dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; rHGH; RNGHGP; Somatotropin |
| Gene Symbols | Gh, Gh1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.