Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Hormone R Antibody (008), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290374MFV610
Description
Growth Hormone R Monoclonal antibody specifically detects Growth Hormone R in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Growth Hormone R | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
008 | |
Immunohistochemistry-Paraffin | |
GH receptor, GHBP, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor | |
Rabbit immunized with purified, recombinant Mouse Growth Hormone R (Accession#: NP_034414.2, Met1-Gln273) | |
0.1 mL | |
Cytokine Research, Signal Transduction | |
2690 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction