Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth hormone receptor Polyclonal Antibody, Bioss
SDP

Catalog No. 502279741
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9741 Supplier Bioss Supplier No. BS23024R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Growth hormone receptor Polyclonal antibody specifically detects Growth hormone receptor in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen Growth hormone receptor
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene GHR
Gene Accession No. P10912
Gene Alias GH receptor; GH-binding protein; GHBP; GHIP; GHR; GHR/BP; growth hormone binding protein; growth hormone receptor; growth hormone receptor precursor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; MGC124963; MGC156665; serum binding protein; Serum-binding protein; Somatotropin receptor; unnamed protein product
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GHR
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2690
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.