Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Growth Hormone Receptor Recombinant Rabbit Monoclonal Antibody (10C11)
GREENER_CHOICE

Catalog No. FEMA549857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549857 Supplier Invitrogen™ Supplier No. MA549857
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Growth Hormone Receptor Recombinant Monoclonal Antibody for ICC/IF, IHC (P), ELISA

Growth hormone receptor (GHR) is a gene located on chromosome 5p13.1, encoding a transmembrane receptor that plays a critical role in mediating the effects of growth hormone, which is crucial for growth, metabolism, and tissue development. GHR is expressed in various tissues, including the liver, muscle, and adipose tissue, and facilitates the growth hormone signaling pathway by binding to growth hormone, leading to receptor dimerization and activation of intracellular signaling cascades such as the JAK2-STAT5 pathway. This signaling promotes growth and anabolic processes, influencing body composition, insulin sensitivity, and lipid metabolism. Mutations or alterations in the GHR gene can lead to growth disorders like Laron syndrome, characterized by insensitivity to growth hormone. Understanding GHR's structure and function is pivotal for developing therapeutic strategies for growth deficiencies and metabolic syndromes, as modifying GHR signaling pathways can offer potential interventions in related conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Growth Hormone Receptor
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10C11
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GHR
Gene Accession No. P10912
Gene Alias GH receptor; GH-binding protein; GHBP; GHIP; GHR; GHR/BP; growth hormone binding protein; growth hormone receptor; growth hormone receptor precursor; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; MGC124963; MGC156665; serum binding protein; Serum-binding protein; Somatotropin receptor; unnamed protein product
Gene Symbols GHR
Host Species Rabbit
Immunogen A synthesized peptide derived from human Growth hormone receptor.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2690
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.