Learn More
Invitrogen™ Growth Hormone Recombinant Rabbit Monoclonal Antibody (JM108-09)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA532604
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Human growth hormone (hGH) is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The GH1 gene, along with 4 additional related genes, is located at the growth hormone locus on chromosome 17. The five genes share a remarkably high degree of sequence identity. Alternative splicing results in 5 isoforms ranging from approximately 19 kDa to 24 kDa. HGH is produced by the somatotroph cells in the pituitary. It acts through binding to membrane receptors that belong to the cytokine receptor subfamily. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues. Mutations or deletions in the gene lead to growth hormone deficiency and short stature.
Specifications
Growth Hormone | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P01241 | |
Gh1 | |
Synthetic peptide within Human Growth Hormone aa 72-106. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
JM108-09 | |
Unconjugated | |
Gh1 | |
dog growth hormone; GH; Gh1; GHB1; GHB5; GHN; GH-N; Growth hormone; growth hormone 1; growth hormone 12; growth hormone B1; growth hormone B5; growth hormone precursor; HGH; hGH-N; IGHD1B; pituitary growth hormone; rHGH; RNGHGP; Somatotropin | |
Rabbit | |
Protein A | |
RUO | |
2688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.