Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120998 Supplier Invitrogen™ Supplier No. PA5120998
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, NCI-H460, Mouse lung, Mouse brain, Mouse stomach, Rat lung, Rat brain, Rat stomach. The target is usually found in the following locations: Secreted. Immunogen sequence: MRGRELPLVL LALVLCLAPR GRAVPLPAGG GTVLTKMYPR GNHWAVGHLM GKKSTGESSS VSERGSLKQQ LREYIRWEEA ARNLLGLIEA KENRNHQPPQ PKALGNQQPS WDSEDSSNFK DVGSKGKVGR LSAPGSQREG RNPQLNQQ.

GRP stimulates gastrin release as well as other gastrointestinal hormones. Operates as a negative feedback regulating fear and established a causal relationship between GRP-receptor gene expression, long-term potentiation, and amygdala-dependent memory for fear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Grp
Gene Accession No. P07492, P24393, Q8R1I2
Gene Alias BLP; BN; bombesin; galectin like; Galectin-like protein; Galectin-related protein; gastrin releasing peptide; gastrin-releasing peptide; GRP; GRP-10; HSPC159; Lectin galactoside-binding-like protein; lectin, galactoside binding like; lectin, galactoside-binding-like; LGALSL; neuromedin C; Neuromedin-C; pre-progastrin releasing peptide; preproGRP; prepro-GRP; proGRP; testicular tissue protein Li 103
Gene Symbols Grp
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human Gastrin Releasing Peptide (GRP) (NP_0020822).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171101, 225642, 2922
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.