Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP75/HSPA9B/Mortalin Antibody (SR2186), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | GRP75/HSPA9B/Mortalin |
---|---|
Clone | SR2186 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
GRP75/HSPA9B/Mortalin Monoclonal antibody specifically detects GRP75/HSPA9B/Mortalin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
GRP75/HSPA9B/Mortalin | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
75 kDa glucose-regulated protein, GRP-75, GRP75mthsp75, Heat shock 70 kDa protein 9, heat shock 70kD protein 9B, heat shock 70kDa protein 9 (mortalin), heat shock 70kDa protein 9B (mortalin-2), HSPA9BMGC4500, Mortalin, mortalin, perinuclear, mortalin-2, MOT, mot-2, MOT2, MTHSP75, p66-mortalin, PBP74CSA, Peptide-binding protein 74, stress-70 protein, mitochondrial | |
A synthesized peptide derived from human GRP75/HSPA9B/Mortalin (Uniprot #: P38646) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2186 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3313 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title