Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP75 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983322
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3322 Supplier Bioss Supplier No. BS1469R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.

Specifications

Antigen GRP75
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HSPA9
Gene Accession No. P38646
Gene Alias 70 kDa heat shock protein; 74kDa; 75 kDa glucose-regulated protein; BOS_8172; C3H-specific antigen; catecholamine-regulated protein 40; CRP40; csa; epididymis secretory sperm binding protein Li 124m; EVPLS; GRP 75; GRP75; GRP-75; Heat shock 70 kDa protein 9; heat shock 70 kDa protein 9B; heat shock 70kD protein 9B; heat shock 70kDa protein 9; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; heat shock 70kDa protein 9B (mortalin-2); Heat shock protein; heat shock protein 9; heat shock protein 9A; heat shock protein cognate 74; heat shock protein family A (Hsp70) member 9; heat shock protein family A (Hsp70) member 9 S homeolog; heat shock protein, 74 kDa, A; heat shock protein, A; HEL-S-124m; Hsc74; HSP; Hsp74; Hsp74a; HSPA9; hspa9.S; Hspa9a; hspa9-a; HSPA9B; hspa9-b; I79_009139; LOC100347135; MGC4500; Mortalin; mortalin, perinuclear; mortalin2; mortalin-2; MOT; MOT2; Mot-2; Mthsp70; mt-HSP70; MTHSP75; p66 MOT; p66-mortalin; PBP74; Peptide-binding protein 74; SAAN; SIDBA4; stress-70 protein mitochondrial-like protein; Stress-70 protein, mitochondrial; Stress-70 protein, mitochondrial-like protein; XELAEV_18020021mg
Gene Symbols HSPA9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GRP75.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.