Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody (4H12), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315138
This item is not returnable.
View return policy
Description
GRP78/HSPA5 Monoclonal antibody specifically detects GRP78/HSPA5 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
GRP78/HSPA5 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
4H12 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
A synthesized peptide derived from human GRP78/HSPA5. UniProt ID (P11021). | |
100 μg | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
3309 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction