Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRP78/HSPA5 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$206.50 - $426.00

Specifications

Antigen GRP78/HSPA5
Concentration 1.0 mg/ml
Dilution Western Blot 1-3 ug/ml, Simple Western 1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Egawa et al (2011)), Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056411T
SDP
View Documents
Novus Biologicals
NB10056411SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056411
SDP
View Documents
Novus Biologicals
NB10056411
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GRP78/HSPA5
Western Blot 1-3 ug/ml, Simple Western 1:250, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Egawa et al (2011)), Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
P11021
3309
A portion of amino acids 550-600 of human GRP78 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.0 mg/ml
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response
PBS with 0.05% Sodium Azide
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2
HSPA5
IgG
Protein G purified
Human and Mouse. Predicted to react with Chimpanzee, Dog, Macaque, Rat, Xenopus and Zebrafish.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.