Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106277R
Description
GRP78/HSPA5 Polyclonal specifically detects GRP78/HSPA5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GRP78/HSPA5 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
Rabbit | |
78 kDa | |
0.1 mL | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response | |
3309 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSPA5 | |
Synthetic peptide made to a C-terminal portion of rat GRP78 (within residues 600-654). [Swiss-Prot# P06761] | |
Affinity Purified | |
RUO | |
Primary | |
Mouse and rat. Reacts weakly with human. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction