Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78/HSPA5 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106274FR
Description
GRP78/HSPA5 Polyclonal antibody specifically detects GRP78/HSPA5 in Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry - Free Floating.Specifications
GRP78/HSPA5 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Free-Floating | |
78 kDa glucose-regulated protein, BiP, Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78, FLJ26106, GRP-78, GRP78BIP, Heat shock 70 kDa protein 5, heat shock 70kD protein 5 (glucose-regulated protein, 78kD), heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa), Immunoglobulin heavy chain-binding protein, MIF2 | |
Rabbit | |
Affinity Purified | |
Cancer, Cellular Markers, ER Markers, Lipid and Metabolism, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
3309 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Free Floating) | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
HSPA5 | |
Synthetic peptide made to an internal portion of human GRP78 (within residues 250-300). [Swiss-Prot# P11021] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, C. elegans, Chicken, Drosophila, Sheep | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction