Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRP78 Monoclonal Antibody (1H11-1H7), PE
GREENER_CHOICE

Catalog No. PIMA545279
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545279 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545279 Supplier Invitrogen™ Supplier No. MA545279
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

0.5 μg/mL of MA5-45279 was sufficient for detection of Grp78 in 10 μg of rat tissue lysate by ECL immunoblot analysis. Detects approximately 78kDa.

GRP78 is a 78 kDa glucose regulated protein that belongs to the family of heat shock proteins that include HSP70. GRP78 is a resident protein of the endoplasmic reticulum (ER) and associates transiently with a variety of newly synthesized secretory and membrane proteins. GRP78 may also interact with mutant and misfolded proteins, marking them for degradation. The highly conserved sequence Lys-Asp-Glu-Leu (KDEL) is present at the C-terminus of GRP78 and other resident ER proteins including glucose regulated protein 94 (GRP94) and protein disulfide isomerase (PDI). The KDEL signal is recognized by the KDEL receptor and this interaction ensures recycling of escaped ER proteins back to the ER. GRP78's involvement in correct folding of proteins and degradation of misfolded proteins is via its interaction with DNAJC10, probably to facilitate the release of DNAJC10 from its substrate. GRP78 is critical for maintenance of cell homeostasis and the prevention of apoptosis. GRP78 protein levels have been shown to be a reliable biomarker of hypoglycemia and serves a neuroprotective function in neurons exposed to glutamate and oxidative stress. GRP78 levels are reduced in the brains of Alzheimer's Disease patients, and decreased expression of GRP78 is found associated with missense mutations in the human presenilin-1 (PS1) gene. Further, the induction of the GRP78 protein has been associated with the development of drug-resistance to antitumor drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRP78
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H11-1H7
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Hspa5
Gene Accession No. P06761, P11021, P20029, Q0VCX2, Q91883
Gene Alias 78 kDa glucose-regulated protein; 78 kDa glucose-regulated protein homolog; 78-kD glucose-regulated protein precursor; AL022860; AU019543; baffled; Binding-immunoglobulin protein; bip; BiP protein; BiP/Grp78; cb865; D2Wsu141e; D2Wsu17e; Endoplasmic reticulum chaperone BiP; endoplasmic reticulum chaperone BiP; 78 kDa glucose-regulated protein; endoplasmic reticulum lumenal Ca(2+)-binding protein grp78; epididymis secretory sperm binding protein Li 89n; fb60h09; fi36d04; FLJ26106; glucose regulated protein, 78 kDa; glucose-regulated protein; glucose-regulated protein, 78kDa; GRP 78; grp78; GRP-78; heat shock 70 kDa protein 5; heat shock 70 kDa protein 5a; heat shock 70kD protein 5; heat shock 70kD protein 5 (glucose-regulated protein, 78kD); heat shock 70kDa protein 5; heat shock 70kDa protein 5 (glucose-regulated protein); heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa); heat shock protein 5; heat shock protein 70 family protein 5; heat shock protein family A (Hsp70) member 5; heat shock protein family A (Hsp70) member 5 L homeolog; heat shock protein family A member 5; heavy-chain binding protein BiP; HEL-S-89n; Hsce70; hsp70; Hsp70 family ATPase KAR2; HSP70 family protein 5; HSPA 5; HSPA5; hspa5.L; hspa5a; I79_019946; immunoglobulin binding protein; immunoglobulin heavy chain-binding protein; Immunoglobulin heavy chain-binding protein homolog; J1248; KAR2; mBiP; MIF2; Sez7; SEZ-7; SSD1; Steroidogenesis-activator polypeptide; wu:fb60h09; wu:fi36d04; XAP-1; XAP-1 antigen; XELAEV_180384511mg; YJL034W; zgc:55994; zgc:77606
Gene Symbols Hspa5
Host Species Mouse
Immunogen His-tagged human GRP78.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008764, 100689305, 14828, 25617, 3309, 397850, 415113
Target Species Human, Mouse, Rat, Rabbit, Monkey, Bovine, Hamster, Frog, Fungi
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.