Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRP78 Mouse anti-Human, Unconjugated, Clone: C19, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS215925UG
Description
GRP78 Monoclonal antibody specifically detects GRP78 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation.
Specifications
GRP78 | |
Monoclonal | |
Unconjugated | |
Endoplasmic reticulum chaperone BiP;EC:3.6.4.10;78 kDa glucose-regulated protein;Binding-immunoglobulin protein;BiP;Heat shock protein 70 family protein 5;HSP70 family protein 5;Heat shock protein family A member 5;Immunoglobulin heavy chain-binding protein | |
Mouse | |
72.33 kDa calculated. | |
25 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
C19 | |
NP_005338 | |
HSPA5, GRP78 | |
1E07 Sca-1+ Cardiac progenitor cells isolated from human heart tissue. | |
Ligatrap | |
Signaling | |
Clone C19 is a mouse monoclonal antibody that specifically detects the glycosylated form of GRP78. Its reactivity is abolished upon deglycosylation. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction