Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRPEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557751
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557751 Supplier Invitrogen™ Supplier No. PA557751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLEKATQCVP KEEIKDDNPH LKNLYEGLVM TEVQIQKVFT KHGLLKLNPV GAKFDPYEHE ALFHTPVEGK EPGTVALVSK VGYK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRPEL1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRPEL1
Gene Accession No. P97576, Q99LP6, Q9HAV7
Gene Alias AA408748; FLJ25609; GREPEL1; GrpE; GrpE like 1, mitochondrial; grpE protein homolog 1, mitochondrial; GrpE#1; GrpE1; GRPEL1; GrpE-like 1; GrpE-like 1, mitochondrial; GrpE-like protein cochaperone; HMGE; mt-GrpE#1; mt-Grpel1; stress-inducible chaperone mt-GrpE#1
Gene Symbols GRPEL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRPEL1. Recombinant protein control fragment (Product #RP-96622).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17713, 79563, 80273
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.