Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRPEL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554723 Supplier Invitrogen™ Supplier No. PA554723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSLWAGRLR VQRLLAWSAA WESKGWPLPF STATQRTAGE DCRSEDPPDE LGPPLAERAL RVKAVKLEKE VQDLTVRYQR AIADCENIRR RTQRCVEDAK IFGIQSFCKD LVE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

GRPEL2 (GrpE Like 2, Mitochondrial) is a Protein Coding gene. Among its related pathways are Mitochondrial protein import and Metabolism of proteins. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL1. Essential component of the PAM complex, a complex required for the translocation of transit peptide-containing proteins from the inner membrane into the mitochondrial matrix in an ATP-dependent manner. Seems to control the nucleotide-dependent binding of mitochondrial HSP70 to substrate proteins. Stimulates ATPase activity of mt-HSP70. May also serve to modulate the interconversion of oligomeric (inactive) and monomeric (active) forms of mt-HSP70.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRPEL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRPEL2
Gene Accession No. O88396, Q8TAA5
Gene Alias actin filament-associated protein 1-like 1; Afap1l1; AFAP1-like protein 1; GrpE like 2, mitochondrial; grpE protein homolog 2, mitochondrial; Grpel2; GrpE-like 2, mitochondrial; Mt-GrpE#2; Mt-grpel2
Gene Symbols GRPEL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRPEL2. Recombinant protein control fragment (Product #RP-91013).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134266, 17714, 688777
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.