Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRSF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569632
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9632 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9632 Supplier Bethyl Laboratories, Inc Supplier No. A305136AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

G-rich sequence factor 1 (GRSF-1) is a cytoplasmic protein that binds RNAs containing the G-rich element. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. GRSF-1 is up-regulated with prostate cancer progression and down regulated by the differentiating agents phenylacetate and phenylbutyrate. The function of GRSF-1 is uncharacterized, but potential roles include mRNA transport, stabilization, or translation. It might react with G-rich containing mRNA of Bcl2 and Bax indicating a regulation role of GRSF-1 in metastatic prostate cancer.

Specifications

Antigen GRSF1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene GRSF1
Gene Accession No. Q12849
Gene Alias G-rich sequence factor 1, GRSF-1
Gene Symbols GRSF1
Host Species Rabbit
Immunogen Between 100 and 150
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2926
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.