Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRTP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101613
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIPA5101613 Supplier Invitrogen™ missing translation for 'supplierNo' PA5101613
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GRTP1.

GRTP1, also known as germ cell-specific gene 1-related protein, is a protein that is expressed in the testis and is involved in the regulation of spermatogenesis, the process of sperm cell development. It is a member of the GRP family of proteins, which are characterized by a conserved motif called the GRP domain. GRTP1 is thought to play a role in the differentiation of spermatogonia into spermatocytes and in the regulation of meiosis, the process of cell division that produces haploid cells. Mutations in the GRTP1 gene have been associated with male infertility. GRTP1 was identified as an HIV dependency factor (HDF), suggesting that GRTP1 may be an important drug target in HIV treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRTP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRTP1
Gene Accession No. Q5TC63
Gene Alias 5430401C05Rik; C81211; GH regulated TBC protein 1; growth hormone regulated TBC protein 1; growth hormone-regulated TBC protein 1; Grtp1; LRRGT00052; RP11-391H12.7; TBC1 domain family member 6; Tbc1d6
Gene Symbols GRTP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRTP1(Accession Q5TC63), corresponding to amino acid residues M79-T129.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79774
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.