Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRWD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584263
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584263 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584263 Supplier Invitrogen™ Supplier No. PA584263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WSPTENTVFA SCSADASIRI WDIRAAPSKA CMLTTATAHD GDVNVISWSR REPFLLSGGD DGALKIWDLR QFKSGSP Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 94%.

This gene encodes a glutamate-rich protein that contains five WD-repeat motifs. The encoded protein may play a critical role in ribosome biogenesis and may also play a role in histone methylation through interactions with CUL4-DDB1 ubiquitin E3 ligase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRWD1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRWD1
Gene Accession No. Q9BQ67
Gene Alias A301; AI504353; CDW4; CUL4- and DDB1-associated WDR protein 4; glutamate rich WD repeat containing 1; glutamate rich WD repeat protein GRWD; glutamate-rich WD repeat containing 1; glutamate-rich WD repeat-containing protein 1; Grwd; GRWD1; KIAA1942; Protein A301; regulator of ribosome biogenesis 1 homolog; RRB1; WD repeat domain 28; WDR28
Gene Symbols GRWD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRWD1. Recombinant protein control fragment (Product #RP-105021).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83743
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.