Learn More
GRWD1/WDR28 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301576A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
GRWD1 is a component of the 50S and 80S preribosomal complexes and plays a role in ribosome biogenesis (Gratenstein et al., 2005 [PubMed 15885502]).Specifications
| GRWD1/WDR28 | |
| Polyclonal | |
| Unconjugated | |
| GRWD1 | |
| CDW4, GRWD, RRB1, WDR28, CUL4- and DDB1-associated WDR protein 4, WD repeat domain 28, glutamate rich WD repeat protein GRWD, glutamate-rich WD repeat-containing protein 1, regulator of ribosome biogenesis 1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83743 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9BQ67 | |
| GRWD1 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.