Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GRXCR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563718 Supplier Invitrogen™ Supplier No. PA563718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSDGKPRKVR FKISSSYSGR VLKQVFEDGQ ELESPKEEYP HSFLQESLET MDGVYGSGEV PRPQMCSPKL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene encodes a protein containing a glutaredoxin domain, which functions in protein S-glutathionylation. A mutation in this gene was found in a family with autoosomal recessive nonsyndromic sensorineural deafness-101.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRXCR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GRXCR2
Gene Accession No. A6NFK2
Gene Alias DFNB101; glutaredoxin and cysteine rich domain containing 2; glutaredoxin domain-containing cysteine-rich protein 1-like protein; Glutaredoxin domain-containing cysteine-rich protein 2; glutaredoxin, cysteine rich 2; GRXCR1-like protein; GRXCR2
Gene Symbols GRXCR2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GRXCR2. Recombinant protein control fragment (Product #RP-100313).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643226
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.