Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GS 143
GREENER_CHOICESDP

Catalog No. 563650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
563650 50 mg
563610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 563650 Supplier Tocris Bioscience™ Supplier No. 5636/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

β-TrCP1 ligase inhibitor

GS 143 is a β-TrCP1 ligase inhibitor. Inhibits IκBα ubiquitination (IC50 = 5.2 μM) without affecting proteasome activity. Blocks LPS-induced expression of inflammatory cytokines in human myelomonocytic cells. Suppresses antigen-induced NFκB expression, inflammation and mucus production in airways of OVA-sensitized mice.

Specifications

Chemical Name or Material 4-[4-[[5-(2-Fluorophenyl)-2-furanyl]methylene]-4,5-dihydro-5-oxo-3-(phenylmethyl)-1H-pyrazol-1-yl]benzoic acid
CAS 916232-21-8
Quantity 50 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C28H19FN2O4
Formula Weight 466.46
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.