Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GS 39783
GREENER_CHOICESDP

Catalog No. 200150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-015-0 50 mg
20-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-015-0 Supplier R&D Systems™ Supplier No. 2001/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of GABAB receptors

Positive allosteric modulator of GABAB receptor function. Potentiates the effects of GABA on [35S]GTPγS binding at recombinant and native GABAB receptors (EC50 values are 2.1 and 3.1μM respectively). Decreases cocaine self-administration, blocks the rewarding properties of nicotine and produces anxiolytic-like activity without the side effects associated with baclofen or benzodiazepines.

Chemical Identifiers

CAS 39069-52-8
Molecular Formula C15H23N5O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 337.44
Purity >98%
Molecular Weight (g/mol) 337.442
Chemical Name or Material GS 39783
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.