Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GS 6201
GREENER_CHOICE

Catalog No. 472710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-271-0 10 mg
47-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-271-0 Supplier Tocris Bioscience™ Supplier No. 4727/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective A2B antagonist

Selective antagonist of adenosine A2B receptors (Ki values are 22, 1070, 1940 and 3280 nM for human A2B, A3, A1 and A2A receptors respectively). Attenuates allergen-, NECA- and AMP-induced airway reactivity in an allergic mouse model of asthma. Also shown to attenuate the inflammatory response during acute myocardial infarction in mice; reduces caspase-1 activity in the heart.

Chemical Identifiers

CAS 752222-83-6
Molecular Formula C21H21F3N6O2

Specifications

Quantity 10 mg
Formula Weight 446.43
Purity >99%
Molecular Weight (g/mol) 446.434
Chemical Name or Material GS 6201
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.