Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551986 Supplier Invitrogen™ Supplier No. PA551986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFTYKFAEEI RRQSFGAVTP GYDFMARLRS GMESILPPSA HELAQNRLHV SITNAKTREN HLVSTFSSRE DLIKVLLASS FVPIYAGLKL VEYKGQKWVD GGLTNALPIL PVGRTVTISP FSGRLDISPQ DKGQLD Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 63%.

This gene encodes a member of the patatin-like family of phospholipases. The encoded enzyme has both triacylglycerol lipase and transacylase activities and may be involved in adipocyte triglyceride homeostasis. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome Y.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PNPLA4
Gene Accession No. P41247
Gene Alias calcium independent phospholipases A2 eta; DXS1283E; GS2; IPLA2 eta; iPLA2eta; patatin like phospholipase domain containing 4; patatin-like phospholipase domain containing 4; patatin-like phospholipase domain-containing protein 4; PNPLA4; Protein GS2
Gene Symbols PNPLA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GS2. Recombinant protein control fragment (Product #RP-92158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.