Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSAP Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178400AF594
Description
GSAP Polyclonal specifically detects GSAP in Human samples. It is validated for Western Blot.Specifications
GSAP | |
Polyclonal | |
Western Blot | |
pigeon homolog (Drosophila) | |
Rabbit | |
0.1 mL | |
54103 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PION | |
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 150-200) [UniProt A4D1B5] | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction