Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSAP Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178376AF647
Description
GSAP Polyclonal specifically detects GSAP in Human samples. It is validated for Western Blot.Specifications
GSAP | |
Polyclonal | |
Western Blot | |
A4D1B5 | |
PION | |
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5] | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
54103 | |
Store at 4C in the dark. |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
pigeon homolog (Drosophila) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction