Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSAP Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1784000.1ML
Description
GSAP Polyclonal specifically detects GSAP in Human samples. It is validated for Western Blot, Simple Western.Specifications
GSAP | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
Gamma-secretase-activating protein, pigeon homolog (Drosophila) | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:1000, Simple Western 1:100 | |
A4D1B5 | |
PION | |
A synthetic peptide made to an internal portion of the human GSAP protein (between residues 150-200) [UniProt A4D1B5] | |
0.1 mL | |
Alzheimers Research, Neurodegeneration, Neuroscience | |
54103 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction