Learn More
GSC Go Science Crazy AC-DC Ammeters
Shop All GSC Products
Click to view available options
:
AC-DC Ammeter
Description
- Range of 0-3A
- Accuracy of ±2.5%
- Includes color-coded binding posts
- Can be used with banana cords
- Full scale and fully enclosed
Specifications
Specifications
| Product Type | AC/DC Ammeter |
| Description | Full scale and fully enclosed |
| For Use With (Application) | Numerous electrical and laboratory experiment |
| Includes | Color coded binding posts, and can be used with banana cord inputs |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov