Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Gas Law Demonstration
SCI_ED

Catalog No. s09586 Shop All GSC Products
Encompass
Click to view available options
:
Gas Law Demonstration

Ideal for teaching the relationship between pressure, temperature and volume.

  • Contains a screw type plunger that creates pressure or vacuum in the cylinder, depending on the direction the plunger is moved
  • A hose is attached from the cylinder to the pressure gauge so you may see the amount of pressure or vacuum being created
  • Thecylinder is equipped with a temperature probe that changes the digital temperature read-out when air is heated from pressure

Specifications

For Use With (Application) Show the relationship between pressure, temperature and volume
Display Type Analog and Digital
Weight (English) 6 lb.
Product Type Gas Law Demonstration
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.