Learn More
GSC Go Science Crazy GSC International Brownlee Electrolysis Apparatus
SCI_ED
Catalog No. S43485A Shop All GSC Products
Click to view available options
Type:
Electrolysis Demonstration Unit
Electrolysis Unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S43485A Electrolysis Unit
S43484A Electrolysis Demonstration Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S43485A Supplier GSC Go Science Crazy Supplier No. 48111
Ideal unit for beginning experiments in electrolysis.
  • Includes J-hooks with platinum tips
  • Acid resistant base
  • Glass battery jar
Specifications
Type Electrolysis Unit
For Use With (Application) For beginning experiments in electrolysis
Includes J-hook electrodes made with an aluminum rod and platinum foil, graduated collection tubes, an acid resistant base, and glass battery jar, instructions
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov